ENA250698434 Enamine

sodium 1,3-thiazole-4-carboxylate

  • MDL number MFCD14705721
  • Cas Number : 1235440-96-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59046

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59046
Availability details for
Quantity
  • Availability
  • Add to Cart