ENA250698435 Enamine

2-(4-ethoxyphenoxy)pyridine-4-carboximidamide hydrochloride

  • MDL number MFCD14705722
  • Cas Number : 1221724-05-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59052

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59052
Availability details for
Quantity
  • Availability
  • Add to Cart