ENA250698437 Enamine

4-[methyl(2-methylpropyl)amino]benzoic acid hydrochloride

  • MDL number MFCD14705725
  • Cas Number : 1221723-09-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59075

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59075
Availability details for
Quantity
  • Availability
  • Add to Cart