ENA250698443 Enamine

1-[4-(2,4-dichlorophenoxy)-3-methylphenyl]methanamine hydrochloride

  • MDL number MFCD14705731
  • Cas Number : 1221724-11-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59128

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59128
Availability details for
Quantity
  • Availability
  • Add to Cart