ENA250698444 Enamine

2-amino-N-(4-hydroxyphenyl)acetamide hydrochloride

  • MDL number MFCD14705732
  • Cas Number : 105396-69-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59133

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59133
Availability details for
Quantity
  • Availability
  • Add to Cart