ENA250698449 Enamine

4-({8-methylimidazo[1,2-a]pyridin-2-yl}methoxy)benzaldehyde

  • MDL number MFCD00725142
  • Cas Number : 1235441-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122...124
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59213

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59213
Availability details for
Quantity
  • Availability
  • Add to Cart