ENA250698452 Enamine

1-(difluoromethyl)-1H-pyrazol-4-amine hydrochloride

  • MDL number MFCD14705743
  • Cas Number : 1221726-31-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 167...169
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59271

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-59271
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart