ENA250698453 Enamine

3-[(pyridin-3-yl)methoxy]benzene-1-carboximidamide trihydrochloride

  • MDL number MFCD14705744
  • Cas Number : 1221726-33-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 3HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59288

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59288
Availability details for
Quantity
  • Availability
  • Add to Cart