ENA250698455 Enamine

3-[(1H-1,3-benzodiazol-1-yl)methyl]benzene-1-carboximidamide trihydrochloride

  • MDL number MFCD14705746
  • Cas Number : 1223492-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 3HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59290

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59290
Availability details for
Quantity
  • Availability
  • Add to Cart