ENA250698463 Enamine

sodium 2-cyano-1-(oxolan-2-yl)eth-1-en-1-olate

  • Cas Number : 1221728-72-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59403

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59403
Availability details for
Quantity
  • Availability
  • Add to Cart