ENA250698465 Enamine

4-(naphthalen-1-yl)-1,2,3,6-tetrahydropyridine hydrochloride

  • MDL number MFCD14705760
  • Cas Number : 100240-18-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-59412

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59412
Availability details for
Quantity
  • Availability
  • Add to Cart