ENA250698467 Enamine

2-(3,5-dimethyl-1H-pyrazol-1-yl)pyridine-4-carboximidamide hydrochloride

  • MDL number MFCD14705763
  • Cas Number : 1221724-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59438

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59438
Availability details for
Quantity
  • Availability
  • Add to Cart