ENA250698469 Enamine

1-[4-(4-fluorophenoxy)-3-methylphenyl]methanamine hydrochloride

  • MDL number MFCD14705765
  • Cas Number : 1235441-44-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59444

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59444
Availability details for
Quantity
  • Availability
  • Add to Cart