ENA250698470 Enamine

methyl(propyl)amine hydrochloride

  • MDL number MFCD14705766
  • Cas Number : 65845-60-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146...148
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59446

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-59446
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart