ENA250698471 Enamine

{5-[3-(3-fluorophenyl)-1,2-oxazol-5-yl]pentyl}(methyl)amine

  • MDL number MFCD14705767
  • Cas Number : 1221723-61-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59463

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59463
Availability details for
Quantity
  • Availability
  • Add to Cart