ENA250698473 Enamine

2-amino-2-(2,4-dimethoxyphenyl)acetonitrile hydrochloride

  • MDL number MFCD14705774
  • Cas Number : 348143-67-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59480

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59480
Availability details for
Quantity
  • Availability
  • Add to Cart