ENA250698500 Enamine

2-(aminomethyl)-N-(propan-2-yl)benzene-1-sulfonamide

  • MDL number MFCD09738060
  • Cas Number : 918865-06-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79...81
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56254

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56254
Availability details for
Quantity
  • Availability
  • Add to Cart