ENA250698501 Enamine

2-methyl-4-(2-methylpiperidine-1-carbonyl)aniline

  • MDL number MFCD09045263
  • Cas Number : 926238-92-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-56260

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56260
Availability details for
Quantity
  • Availability
  • Add to Cart