ENA250698506 Enamine

3-{[(4-methylpentyl)oxy]methyl}aniline

  • MDL number MFCD11196364
  • Cas Number : 1039947-92-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56305

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56305
Availability details for
Quantity
  • Availability
  • Add to Cart