ENA250698508 Enamine

3-[(4-methoxyphenyl)amino]-1$l^{6}-thiolane-1,1-dione

  • MDL number MFCD00721065
  • Cas Number : 303992-20-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109...111
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56345

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56345
Availability details for
Quantity
  • Availability
  • Add to Cart