ENA250698509 Enamine

5-[4-(propan-2-yl)phenyl]-1H-1,2,3,4-tetrazole

  • MDL number MFCD03069379
  • Cas Number : 460363-75-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 186...188
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-17748

Safety Information

msds https://www.enaminestore.com/catalog/EN300-17748
Availability details for
Quantity
  • Availability
  • Add to Cart