ENA250698511 Enamine

N-(5-amino-2-methylphenyl)pyridine-4-carboxamide

  • MDL number MFCD02585699
  • Cas Number : 436089-25-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-56385

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56385
Availability details for
Quantity
  • Availability
  • Add to Cart