ENA250698513 Enamine

trimethyl({2-phenylimidazo[1,2-a]pyridin-3-yl}methyl)azanium iodide

  • MDL number MFCD07290464
  • Cas Number : 365213-50-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data I-
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-18492

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-18492
Availability details for
Quantity
  • Availability
  • Add to Cart