ENA250698521 Enamine

diethyl(2-phenoxyethyl)amine hydrochloride

  • MDL number MFCD00828594
  • Cas Number : 21663-15-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-18651

Safety Information

msds https://www.enaminestore.com/catalog/EN300-18651
Availability details for
Quantity
  • Availability
  • Add to Cart