ENA250698525 Enamine

4-[bis(2-chloroethyl)amino]benzonitrile

  • MDL number MFCD00181241
  • Cas Number : 71601-35-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-18667

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-18667
Availability details for
Quantity
  • Availability
  • Add to Cart