ENA250698529 Enamine

3-[(cyclohexyloxy)methyl]aniline

  • MDL number MFCD09815245
  • Cas Number : 782405-31-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56486

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56486
Availability details for
Quantity
  • Availability
  • Add to Cart