ENA250698530 Enamine

2-phenylpropan-2-amine

  • MDL number MFCD00134680
  • Cas Number : 585-32-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-56487

Safety Information

signalword Danger
pcodes P210,P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P370+P378,P403+P235,P405,P501
hcodes C
MSDS https://www.enaminestore.com/catalog/EN300-56487
pictograms GHS05
hstatements H227,H314,H318
Availability details for
Quantity
  • Availability
  • Add to Cart