ENA250698532 Enamine

1-benzyl-3-(pyrrolidin-2-yl)piperidine

  • MDL number MFCD05189289
  • Cas Number : 524674-29-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56490

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56490
Availability details for
Quantity
  • Availability
  • Add to Cart