ENA250698535 Enamine

4,4,4-trifluoro-3-hydroxybutanenitrile

  • MDL number MFCD11099999
  • Cas Number : 57165-85-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56506

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56506
Availability details for
Quantity
  • Availability
  • Add to Cart