ENA250698540 Enamine

2-(3-fluorophenyl)-1-(piperazin-1-yl)ethan-1-one

  • MDL number MFCD08442094
  • Cas Number : 926226-12-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56529

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56529
Availability details for
Quantity
  • Availability
  • Add to Cart