ENA250698546 Enamine

4-(4-ethylpiperazin-1-yl)-3-fluorobenzonitrile

  • MDL number MFCD11038744
  • Cas Number : 1157055-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56549

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56549
Availability details for
Quantity
  • Availability
  • Add to Cart