ENA250698548 Enamine

(2-phenyl-1,3-oxazol-4-yl)methanol

  • MDL number MFCD06738534
  • Cas Number : 59398-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79...81
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56551

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56551
Availability details for
Quantity
  • Availability
  • Add to Cart