ENA250698555 Enamine

ethyl 4-(2,6-dichloro-3-fluorophenyl)-2,4-dioxobutanoate

  • MDL number MFCD12548642
  • Cas Number : 1041553-00-7
  • Molecular Weight : 307.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56559

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56559
Availability details for
Quantity
  • Availability
  • Add to Cart