ENA250698575 Enamine

2-butylpropanedinitrile

  • MDL number MFCD06215024
  • Cas Number : 7391-58-4
  • Molecular Weight : 122.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-19937

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-19937
Availability details for
Quantity
  • Availability
  • Add to Cart