ENA250698581 Enamine

8,9-dichloro-2,3,4,5-tetrahydro-1-benzoxepin-5-one

  • MDL number MFCD11207524
  • Cas Number : 1094253-83-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103...105
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56867

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56867
Availability details for
Quantity
  • Availability
  • Add to Cart