ENA250698589 Enamine

2-[(6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolin-2-yl)methyl]aniline

  • MDL number MFCD00874309
  • Cas Number : 114688-61-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56876

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56876
Availability details for
Quantity
  • Availability
  • Add to Cart