ENA250698591 Enamine

[2-(dimethylamino)ethyl](3-phenylpropyl)amine

  • MDL number MFCD11154684
  • Cas Number : 101507-01-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-56879

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56879
Availability details for
Quantity
  • Availability
  • Add to Cart