ENA250698598 Enamine

1-(3-amino-2-methylpropyl)pyrrolidin-2-one

  • MDL number MFCD11653480
  • Cas Number : 1099666-77-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56887

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56887
Availability details for
Quantity
  • Availability
  • Add to Cart