ENA250698610 Enamine

2-(2,4-dichlorophenyl)-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD07376332
  • Cas Number : 1094355-53-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56903

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56903
Availability details for
Quantity
  • Availability
  • Add to Cart