ENA250698621 Enamine

2-bromo-3-methyl-N-(pyridin-3-ylmethyl)butanamide

  • MDL number MFCD11119124
  • Cas Number : 1036540-46-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56914

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56914
Availability details for
Quantity
  • Availability
  • Add to Cart