ENA250698624 Enamine

3-[(pyrimidin-2-ylsulfanyl)methyl]-1-benzofuran-2-carboxylic acid

  • MDL number MFCD13358734
  • Cas Number : 1039812-24-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56917

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56917
Availability details for
Quantity
  • Availability
  • Add to Cart