ENA250698627 Enamine

3-{[(4-fluorophenyl)sulfanyl]methyl}-1-benzofuran-2-carboxylic acid

  • MDL number MFCD13378778
  • Cas Number : 1040084-06-7
  • Molecular Weight : 302.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-56920

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56920
Availability details for
Quantity
  • Availability
  • Add to Cart