ENA250698630 Enamine

3-{[(4-hydroxypyrimidin-2-yl)sulfanyl]methyl}-1-benzofuran-2-carboxylic acid

  • MDL number MFCD13383585
  • Cas Number : 1039922-41-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56923

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56923
Availability details for
Quantity
  • Availability
  • Add to Cart