ENA250698638 Enamine

2-(benzyloxy)-N'-hydroxybenzene-1-carboximidamide

  • Cas Number : 936497-77-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-56945

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56945
Availability details for
Quantity
  • Availability
  • Add to Cart