ENA250698639 Enamine

6-(pyridin-2-ylsulfanyl)pyridin-3-amine

  • MDL number MFCD00829041
  • Cas Number : 247060-76-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56948

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56948
Availability details for
Quantity
  • Availability
  • Add to Cart