ENA250698640 Enamine

6-amino-1-[(4-chlorophenyl)methyl]-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD06260079
  • Cas Number : 179486-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-56951

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56951
Availability details for
Quantity
  • Availability
  • Add to Cart