ENA250698644 Enamine

[(4-fluorophenyl)methyl](prop-2-yn-1-yl)amine hydrochloride

  • MDL number MFCD07170806
  • Cas Number : 1158359-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56956

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56956
Availability details for
Quantity
  • Availability
  • Add to Cart