ENA250698645 Enamine

N-(2-aminoethyl)cyclopropanecarboxamide

  • MDL number MFCD09927906
  • Cas Number : 53673-05-5
  • Molecular Weight : 128.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-22410

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-22410
pictograms GHS05,GHS07
hstatements H302,H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart