ENA250698646 Enamine

5-methyl-4-sulfanylidene-1H,4H-thieno[2,3-d]pyrimidine-6-carboxylic acid

  • MDL number MFCD14705551
  • Cas Number : 1176120-44-7
  • Molecular Weight : 226.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-22893

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-22893
Availability details for
Quantity
  • Availability
  • Add to Cart