ENA250698649 Enamine

3-(3,5-dichlorophenyl)-3-oxopropanenitrile

  • MDL number MFCD00662057
  • Cas Number : 69316-09-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56961

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56961
Availability details for
Quantity
  • Availability
  • Add to Cart